Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCCS Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526880

Catalog No. PIPA526880

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is an enzyme that covalently links a heme group to the apoprotein of cytochrome c. Defects in this gene are a cause of microphthalmia syndromic type 7 (MCOPS7). Three transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

HCCS
Polyclonal
PBS with 0.09% sodium azide
P53701, P53702
CCHL, MCOPS7, cytochrome c heme-lyase, cytochrome c-type heme lyase, holocytochrome c-type synthase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
15159, 3052
Immunohistochemistry, Western Blot
Unconjugated
HCCS
Liquid
HCCS
KLH conjugated synthetic peptide between 173-200 amino acids from the central region of human HCCS
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit