Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523823

Catalog No. PIPA523823


Add to cart

Description

Description

This antibody also recognizes isoform 2 of PCDP1, which has a predicted molecular weight of ~64.2KDa.

Specifications

Specifications

PCDP1
Polyclonal
PBS with 0.09% sodium azide
Q4G0U5, A9Q751
hCG_17324
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
200373, 226356
Flow Cytometry, Western Blot
Unconjugated
CFAP221
Liquid
CFAP221
KLH conjugated synthetic peptide between 787-816 amino acids from the C-terminal region of human PCDP1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit