Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISPD Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525854

Catalog No. PIPA525854


Add to cart

Specifications

Specifications

ISPD
Polyclonal
PBS with 0.09% sodium azide
A4D126, Q5RJG7
Nip, 2-C-methyl-D-erythritol 4-phosphate cytidylyltransferase-like protein, 4-diphosphocytidyl-2C-methyl-D-erythritol synthase homolog, isoprenoid synthase domain-containing protein, notch1-induced protein, hCG_1745121
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
729920, 75847
Western Blot
Unconjugated
ISPD
Liquid
ISPD
KLH conjugated synthetic peptide between 400-429 amino acids from the C-terminal region of human ISPD
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit