Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHISA7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525713

Catalog No. PIPA525713

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SHISA7
Polyclonal
Unconjugated
SHISA7
Liquid
SHISA7
KLH conjugated synthetic peptide between 308-337 amino acids from the central region of human SHISA7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
A6NL88, Q8C3Q5
UPF0626 protein A, protein shisa-7, hCG_2039146
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
232813, 729956
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit