Learn More
hCG beta, clone: HCG-60, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA119063
Catalog No. 01-675-036
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
hCG is a protein of the molecular weight about 40 kDa. hCG belongs to the glycoprotein hormone family together with lutropin (LH), follitropin (FSH) and thyrotropin (TSH). hCG is synthesised by trophoblastic cells of the placenta during pregnancy and stimulates the growth of corpus luteum. The other hormones are produced by anterior pituitary gland. All these hormones are structurally related being composed of two noncovalently associated subunits alpha and beta.
Specifications
Specifications
hCG beta
Monoclonal
0.5mg/mL
PBS with 15mM sodium azide; pH 7.4
P0DN86
Chorionic Gonadotropin
Murine
IgG1
100μg
4° C, do not freeze, do not freeze
Primary
1082
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin)
HCG-60
Unconjugated
CGB3
Liquid
CGB3
Human chorionic gonadotropin.
Protein A
RUO
Antibody
Monoclonal
Human