Learn More
Hck Mouse, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 610278
 View more versions of this product
Catalog No. BDB610278
Add to cart
Description
Description
The hematopoietic cell kinase (Hck), a member of the Src family of tyrosine kinases, is primarily expressed in cells of the hematopoietic system. Hck levels have been reported to increase with terminal differentiation in monocyte/macrophages and granulocytes and is further elevated during macrophage activation. Hck also has been reported to be expressed in undifferentiated embryonic stem (ES) cells. This expression is substantially reduced when ES cells are allowed to differentiate in vitro by withdrawal of leukemia inhibitory factor (LIF). Like that of other Src family tyrosine kinases, Hck activity is downregulated by phosphorylation of a conserved C-terminal tyrosine residue. This has reportedly been examined through a “hit and run” gene targeting strategy where Hck tyrosine kinase activity was elevated by replacing the Hck tyrosine 499 residue with phenylalanine. Results indicated that LIF receptor (LIFR) signaling is partially mediated by Hck. Furthermore, Hck is contained within the LIFR complex and is physically associated with gp130, the common signal transducing component of functional LIFR and IL-6 receptor complexes.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Specifications
Hck
18
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Monoclonal
Immunofluorescence, Immunoprecipitation, Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Primary
Canine, Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.