Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCNK1 Mouse anti-Human, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611734

Catalog No. BDB611734


Add to cart

Description

Description

Proteins of the Ras superfamily play critical roles in the control of normal and neoplastic proliferation. These proteins relay signals from Tyr-kinases at the plasma membrane to the nucleus via a network of Ser/Thr kinases that includes the MAP kinase (Raf-MEK-ERK) pathway. Kinase suppressor of Ras (KSR-1) was discovered in Drosophila in a genetic screen that identified mutations that suppress constitutively active Ras mutants. Connector enhancer of KSR (CNK) was found in a similar screen to identify mutations that enhance the KSR- dependent phenotype. Drosophila CNK contains a sterile alpha motif (SAM), a conserved region in CNK (CRIC), and a PDZ domain in the N-terminal region, a proline-rich and a plecstrin homology (PH) domain in the central region, and a C-terminal Pro-rich domain. The human homologue of CNK (hCNK1) contains similar N-terminal and central domains, but is 713 amino acids in length compared to 1557 amino acids for Drosophila CNK. In Drosophila, the N-terminal region of CNK facilitates binding to RAS, while the C-terminal region inhibits RAS- and RAF-dependent signaling. Thus, CNK may be important for regulation of both RAS- and RAF-dependent signaling.

Host Species: Mouse
Clone: 46
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human hCNK1 aa. 10-217

Immunofluorescence, Western Blotting

Specifications

Specifications

hCNK1
46
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Connector enhancer of KSR
Affinity Purified
Human hCNK1 aa. 10-217
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit