Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HDAC1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP216788

Catalog No. NBP216788

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HDAC1 Polyclonal antibody specifically detects HDAC1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HDAC1
1mg/mL
Western Blot 1:500-1:3000
Q13547
EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human. Expected to cross react based on sequence identity: Mouse (100%), Chicken (98%), Rat (100%), Zebrafish (97%), Bovine (100%), Xenopus tropicalis (97%).
Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
HDAC1
Recombinant protein encompassing a sequence within the center region of human HDAC1. The exact sequence is proprietary.
55 kDa
0.1 ml
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Histone Deacetylases, Stem Cell Markers
Primary
3065
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only