Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-HDAC2, Clone: Y461, ™ (Upstate™) Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 04229/DEL

Catalog No. 50-171-308

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly conserved from yeast to human.

Host Species:Rabbit
Clone:Y461
Isotype:IgG
Species Reactivity:Human, Mouse, Rat
Epitope:C-terminus
Immunogen:KLH-conjugated to a synthetic peptide from the C-terminal region of human Histone Deacetylase 2.
Molecular Weight:55kDa

Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Specifications

Specifications

HDAC2
Supernatant
IgG
RUO
Primary
Human, Murine, Rat
Unconjugated
Rabbit
100μL
Apoptosis, Epigenetics
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit