Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HDAC6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178981

 View more versions of this product

Catalog No. NBP178981


Add to cart

Description

Description

HDAC6 Polyclonal antibody specifically detects HDAC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot (Negative), Immunohistochemistry (Negative).
Specifications

Specifications

HDAC6
1.07 mg/mL
Flow Cytometry, Immunocytochemistry/Immunofluorescence 1:100, Western Blot (Negative), Immunohistochemistry (Negative)
Q9UBN7
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Rabbit
IgG
0.1 ml
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Neuroscience
Primary
10013
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry
Unconjugated
PBS, 30% Glycerol with 0.05% Sodium Azide
Affinity Purified
HDAC6
A synthetic peptide made to a C-terminal portion of the human HDAC6 protein (between residues 1175-1215). [UniProt Q9UBN7]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human and mouse. Mouse data from customer feedback.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only