Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HDAC6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7]
Antigen HDAC6
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056343T
View Documents
Novus Biologicals
NB10056343SS
0.025 mg
Each for $119.60
Add to cart
 
NB10056343
View Documents
Novus Biologicals
NB10056343
0.1 mg
Each for $314.00
Add to cart
 
Description

Description

HDAC6 Polyclonal antibody specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunoprecipitation.
Specifications

Specifications

Rabbit
Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7]
Polyclonal
Primary
Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
NP_002495
PBS with 0.02% Sodium Azide
IgG
HDAC6
Purified
RUO
In NIH 3T3, a 134 kDa band is observed.
10013
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit