Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HDAC7 (p Ser318), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP203978SS

 View more versions of this product

Catalog No. NBP203978SS


Add to cart

Description

Description

HDAC7 p Ser318 Polyclonal antibody specifically detects HDAC7 p Ser318 in Human, Bovine, Canine, Equine, Primate, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

HDAC7 (p Ser318)
1.0 mg/mL
Western Blot 0.1 - 1 ug/mL
NP_055522
DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917
Rabbit
IgG
0.025 mg
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific
Primary
51564
Human, Bovine, Canine, Equine, Primate, Zebrafish
Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
HDAC7
A synthetic peptide containing phospho-serine 318 of human HDAC7 was used as the immunogen.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reacts with Chimpanzee, Cow, Dog, Horse, Human, New World Monkey and Zebrafish. Predicted to react with Mouse, Rat and Chicken.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only