Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatoma-derived growth factor, related protein 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human HDGFRP3 protein.

Manufacturer:  Abnova Corporation H00050810B01

Catalog No. 89-007-551


Add to cart

Description

Description

Sequence: MARPRPREYKAGDLVFAKMKGYPHWPARIDELPEGAVKPPANKYPIFFFGTHETAFLGPKDLFPYKEYKDKFGKSNKRKGFNEGLWEIENNPGVKFTGYQAIQQQSSSETEGEGGNTADASSEEEGDRVEEDGKGKRKNEKAGSKRKKSYTSKKSSKQSRKSPGDEDDKDCKEEENKSSSEGGDAGNDTRNTTSDLQKTSEGT
Specifications

Specifications

hepatoma-derived growth factor, related protein 3
Unconjugated
No additive
NM_016073.2
CGI-142|HDGF2
Mouse
50 uL
Cytokines
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human HDGFRP3 protein.
HDGFRP3
Serum
HDGFRP3
HDGFRP3 (NP_057157.1, 1 a.a. ~ 203 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
50810
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit