Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoA1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129508

Catalog No. PIPA129508

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.

PA1-29508 detects HDL from human samples.PA1-29508 has been successfully used in Western blot applications.The PA1-29508 immunogen is: Full length native plasma protein (purified) (Human).
Specifications

Specifications

HDL
Polyclonal
PBS with 0.01% sodium azide
P02647
11-DH, 11-beta-HSD1, HDL, HSD11, HSD11B, HSD11L, MGC13539, SDR26C1
Chicken
IgY
50μg
Store at -20°C
Primary
335
Western Blot
Unconjugated
APOA1
Purified
APOA1
Full length native plasma protein (purified) (Human).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit