Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDLBP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526422

Catalog No. PIPA526422

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
Specifications

Specifications

HDLBP
Polyclonal
PBS with 0.09% sodium azide
Q00341
HBP, PRO2900, VGL, HDL-binding protein, high density lipoprotein-binding protein, vigilin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3069
Immunohistochemistry, Western Blot
Unconjugated
HDLBP
Liquid
HDLBP
KLH conjugated synthetic peptide between 764-791 amino acids from the central region of human HDLBP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit