Learn More
Thermo Scientific Lab Vision Heat Shock Factor 1 (HSF1) Ab-1, Rat Monoclonal Antibody
Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments.
Manufacturer: Thermo Scientific RT405P1
Description
Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments with Thermo Scientific Heat Shock Factor 1 (HSF1) Ab-1, Rat Monoclonal Antibody.
HSF1 responds to cellular stress signals such as heat, heavy metals, and oxidative reagents; binds to heat shock response elements (HSEs) in the upstream region of heat shock gene and activates their transcription. Some laboratories have reported that the constitutive, non-DNA binding, monomeric form of inactive HSF1 is located in the cytoplasm. Upon activation, HSF1 is trimerized and is localized in nucleus where it binds DNA.Host Species: Rat
Clone: 4B4
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 425-439 of mouse HSF1
Immunogen: Recombinant mouse HSF1 protein (aa 1-503)
Molecular Weight: 70-85 kDa (depending upon the source and state of cells)
Positive Control: LS174T or MAD109 cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Heat Shock Factor 1 (HSF1) Ab-1 | |
Monoclonal | |
200µg/mL | |
Concentrated | |
Recombinant mouse HSF1 protein (aa1-503) | |
500μL | |
Stress Response | |
Primary | |
Human, Murine |
Immunoprecipitation, Western Blot | |
4B4 | |
Unconjugated | |
Rat | |
IgG1 | |
RUO | |
Primary | |
Monoclonal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.