Learn More
HEC Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 611041
 View more versions of this product
Catalog No. BDB611041
Add to cart
Description
Description
Cell division is a highly regulated process which ensures that properly replicated DNA is received by both daughter cells. During M phase, the pairs of sister chromatids separate into each daughter cell. To ensure correct cell division, each step of this process is highly regulated by proteins such as kinases and phosphatases that serve as checkpoints. HEC (highly expressed in cancer) was discovered during a search for mitotic proteins that might provide insight to the progression of malignant cells. HEC was identified via its interaction with Rb (retinoblastoma protein) in the yeast two hybrid system. Its expression is elevated during M phase, and it localizes to the centromeres during this time of rapid division. During G1 HEC is dispersed in the nucleus. In addition, HEC is abundant in rapidly dividing cancer cells, but is absent from terminally differentiated cells. Also, microinjection of HEC antibodies resulted in abnormal chromosome distribution. Therefore, HEC is thought to play a role in chromosome segregation during the M phase of the cell cycle.
Immunofluorescence, Western Blotting
Specifications
Specifications
HEC
1
Unconjugated
Mouse
IgG1
RUO
Primary
Canine, Human
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human HEC aa. 495-608
150μg
Cell Biology
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.