Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter pylori, clone: 51-13, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135390

Catalog No. 01-671-445

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.

Specifications

Specifications

Helicobacter pylori
Monoclonal
Unconjugated
Liquid
Murine
IgG1
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence
51-13
ascites diluted in PBS with 1% BSA and 0.1% sodium azide
H. pylori, Campylobacter pyloridis
Helicobacter pylori
1mL
4° C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit