Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Helios, Clone: 22F6, BioLegend™ Non-distribution product as customer accommodation.

Manufacturer:  BiolegendSupplier Diversity Partner 137216

Catalog No. NC0108895


Add to cart

Description

Description

Host Species: Hamster
Clone: 22F6
Isotype: IgG
Cross Reactivity: Human, Mouse
Immunogen: Helios peptide

Flow Cytometry

Specifications

Specifications

Helios
PE
Hamster
IgG
Primary
Human, Murine
22F6
Unpurified
Helios peptide
100 Tests
Monoclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit