Learn More
HELLS Mouse anti-Human, Clone: 1D10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519067
Catalog No. PIMA519067
Add to cart
Description
Description
Peptide Sequence: MFGSSEKETI ELSPTGRPKR RTRKSINYSK IDDFPNELEK LISQIQPEVD RERAVVEVNI PVESEVNLKL QNIMMLLRKC CNHPYLIEYP IDPVTQEFKI DEELVTNSGK FLILDRMLPE LKKRGHKVLL FSQMTSMLDI LMDYCHLRDF NFSRLDGSMS YSEREKNMHS FNTDPEVFIF LVSTRAGGLG INLTAADTVI IYDSDWNPQS DLQAQDRCHR IGQTKPVVVY RLVTANTIDQ KIVERAAAKR KLEKLIIHKN HFKGGQSGLN LSKNFLDPKE LMELLKSRDY EREIKGSREK VISDKDLELL LDRSDLIDQM NASGPIKEKM GIFKILENSE DSSPECLF
This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
Specifications
HELLS
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9NRZ9
LSH, PASG, SMARCA6, SWI/SNF2-related matrix-associated actin-dependent regulator of chromatin subfamily A member 6, SWI/SNF2-related, matrix-associated, actin-dependent regulator of chromatin, subfamily A, member 6, lymphoid-specific helicase, proliferation-associated SNF2-like
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3070
ELISA, Western Blot
1D10
Unconjugated
HELLS
Liquid
HELLS
HELLS (AAH30963, 1 a.a. ∽ 348 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human