Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

helicase, lymphoid-specific, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human HELLS protein.

Manufacturer:  Abnova Corporation H00003070B01

Catalog No. 89-005-516


Add to cart

Description

Description

This gene encodes a lymphoid-specific helicase. Other helicases function in processes involving DNA strand separation, including replication, repair, recombination, and transcription. This protein is thought to be involved with cellular proliferation and may play a role in leukemogenesis. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Sequence: MPAERPAGSGGSEAPAMVEQLDTAVITPAMLEEEEQLEAAGLERERKMLEKARMSWDRESTEIRYRRLQHLLEKSNIYSKFLLTKMEQQQLEEQKKKEKLERKKESLKVKKGKNSIDASEEKPVMRKKRGREDESYNISEVMSKEEILSVAKKNKKENEDENSSSTNLCVEDLQKNKDSNSIIKDRLSETVRQNTKFFFDPVRKCNGQPVPFQQPKHFTGGVMRWYQVEGMEWLRMLWENGINGILADEMGLGKTVQCIATIALMIQRGVPGPFLVCGPLSTLPNWMAEFKRFTPDIPTMLYHGTQEERQKLVRNIYKRKGTLQIHPVVITSFEIAMRDRNALQHCYWKYLIVDEGHRIKNMKCRLIRELKRFNADNKLLLTGTPLQNNLSELWSLLNFLLPDVFDDLKSFESWFDITSLSETAEDIIAKEREQNVLHMLHQILTPFLLRRLKSDVALEVPPKREVVVYAPLSKKQEIFYTAIVNRTIANMFGSSEKETIELSPTGRPKRRTRKSINYSKIDDFPNELEKLISQIQPEVDRERAVVEVNIPVESEVNLKLQNIMMLLRKCCNHPYLIEYPIDPVTQEFKIDEELVTNSGKFLILDRMLPELKKRGHKVLLFSQMTSMLDILMDYCHLRDFNFSRLDGSMSYSEREKNMHSFNTDPEVFIFLVSTRAGGLGINLTAADTVIIYDSDWNPQSDLQAQDRCHRIGQTKPVVVYRLVTANTIDQKIVERAAAKRKLEKLIIHKNHFKGGQSGLNLSKNFLDPKELMELLKSRDYEREIKGSREKVISDKDLELLLDRSDLIDQMNASGPIKEKMGIFKILENSEDSSPECLF
Specifications

Specifications

helicase, lymphoid-specific
Unconjugated
No additive
NM_018063
FLJ10339|LSH|Nbla10143|PASG|SMARCA6
Mouse
50 uL
Stem Cell Biology
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human HELLS protein.
HELLS
Serum
HELLS
HELLS (NP_060533, 1 a.a. ~ 838 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3070
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit