Learn More
Helospectin Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136088
Catalog No. 01-672-265
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
O-linked and free exendin-1 and exendin-1b (Helospectin) have vasoactive intestinal peptide(VIP)/secretin-like biological activities. They interact with rat and human VIP receptors 1 (VIPR1) and 2 (VIPR2), with the highest affinity for the human VIPR2. They induce hypotension that is mediated by relaxation of cardiac smooth muscle.
Specifications
Specifications
Helospectin
Polyclonal
whole serum with 0.1% sodium azide
Exendin-1; Helospectin I; Helospectin-1; Exendin-1b; Helospectin II; Helospectin-2
Synthetic helospectin I.
50 μL
4° C
Primary
Mouse, Rat
Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Lyophilized
Rabbit
IgG
RUO
Antibody
Polyclonal