Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEM1 Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179231

Catalog No. NBP179231


Add to cart

Description

Description

HEM1 Polyclonal antibody specifically detects HEM1 in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

HEM1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
NCKAP1L
Synthetic peptide directed towards the C terminal of human NCKAP1L. Peptide sequence LPLLATDPSSFYSIEKDGYNNNIHCLTKAIIQVSAALFTLYNKNIETHLK.
Immunogen affinity purified
RUO
Primary
3071
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_005328
Hematopoietic protein 1HEM1Membrane-associated protein HEM-1, NCK-associated protein 1-like
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Guinea pig: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Mouse: 92%; Rat: 92%; Canine: 85%; Bovine: 84%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only