Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HEM1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128474

Catalog No. NBP128474


Add to cart

Description

Description

HEM1 Polyclonal antibody specifically detects HEM1 in Human, Mouse, Rat, Canine, Equine samples. It is validated for Peptide ELISA.
Specifications

Specifications

HEM1
0.5 mg/mL
Peptide ELISA Detection limit 1:16000
NP_005328.2
Hematopoietic protein 1HEM1Membrane-associated protein HEM-1, NCK-associated protein 1-like
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Canine, Equine, Human, Mouse, Rat
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NCKAP1L
Peptide with sequence C-RHTENVTKTKTPE corresponding to internal region according to NP_005328.2.
Immunogen affinity purified
RUO
Primary
3071
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only