Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEMK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524489

Catalog No. PIPA524489

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable protein methyltransferase. May methylate a Gln residue in target proteins.
Specifications

Specifications

HEMK1
Polyclonal
PBS with 0.09% sodium azide
Q9Y5R4
HEMK, MTQ1, HEMK homolog 7kb, hemK methyltransferase family member 1, m.HsaHemKP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51409
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HEMK1
Liquid
HEMK1
KLH conjugated synthetic peptide between 311-338 amino acids from the C-terminal region of human HEMK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit