Learn More
Hepatitis B Virus Core Antigen Mouse anti-Virus, Clone: 10E11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA17608
Catalog No. 01-670-342
Add to cart
Description
Description
MA1-7608 detects HBcAg from virus samples. MA1-7608 has been successfully used in ELISA, Western blot, immunoprecipitation and immunohistochemistry procedures. The MA1-7608 immunogen is a purified recombit HBcAg core protein. The epitope corresponds to residues 1-10. Store short-term at 4°C, long-term at -80°C.
The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. Hepatitis B virus is spherical in shape with a diameter of 42nm. It contains a 27 nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
Specifications
Specifications
Hepatitis B Virus Core Antigen
Monoclonal
Unconjugated
Liquid
Mouse
IgG1
RUO
Antibody
Monoclonal
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
10E11
PBS with 0.01% sodium azide
HepB; Hepatitis B virus nucleocapsid particles; HBcAg
Purified recombit HBcAg.
100 μg
4°C or -80°C if preferred
Primary
Virus