Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus Surface Ad/Ay Polyclonal Antibody, Biotin, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA173084

Catalog No. PA173084


Add to Cart

Description

Description

The PA1-73084 antibody reacts with Hepatitis B surface antigen Ad/Ay. PA1-73084 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73084 antibody was raised against surface antigen subtypes ad & ay.

Hepatitis B Virus Surface Ad/Ay Polyclonal antibody specifically detects Hepatitis B Virus Surface Ad/Ay in Virus samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hepatitis B Virus Surface Ad/Ay
Polyclonal
Biotin
HBsAg; HBV; hepatitis b virus surface antigen; HepB
Surface antigen subtypes ad & ay.
1 mL
Primary
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Liquid
ELISA, Immunohistochemistry, Western Blot
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Goat
purified
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit