Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B Virus Surface Ad/Ay Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA173088
Description
The PA1-73088 antibody reacts with Hepatitis B surface antigen Ad/Ay. PA1-73088 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73088 antibody was raised against surface antigen subtypes ad & ay. Recommended short term (less than 6 months) storage is at 4oC. For long term storage, aliquot and store at -20oC, avoiding freeze/thaw cycles.
Hepatitis B Virus Surface Ad/Ay Polyclonal antibody specifically detects Hepatitis B Virus Surface Ad/Ay in Virus samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Hepatitis B Virus Surface Ad/Ay | |
Polyclonal | |
Biotin | |
HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
Surface antigen subtypes ad & ay. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction