Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B Virus Surface Monoclonal Antibody (S 38), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA17604
Description
MA1-7604 detects HBsAg from virus samples. MA1-7604 has been successfully used in ELISA, Immunohistochemistry procedures. The MA1-7604 immunogen is HBsAg purified from human sera pool.
Hepatitis B Virus Surface Monoclonal antibody specifically detects Hepatitis B Virus Surface in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy.Specifications
Hepatitis B Virus Surface | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.01% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy | |
S 38 | |
Unconjugated | |
HBsAg; HBV; hepatitis b virus surface antigen; HepB | |
HBsAg purified from human sera pool. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction