Learn More
Hepatitis C Virus NS4 Mouse anti-Virus, Clone: 1G7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA17593
Catalog No. 01-670-327
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-7593 detects HCV from virus samples. MA1-7593 has been successfully used in ELISA, Western blot, flow cytometry, immunoprecipitation and immunohistochemistry procedures. The MA1-7593 immunogen is a purified recombit HCV NS4 protein.
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Specifications
Specifications
Hepatitis C Virus NS4
Monoclonal
1 mg/mL
PBS with 0.01% sodium azide; pH 7.2
HepC; HCV; HepC NS4; HCV NS4
Purified recombit HCV NS4 protein.
Affinity chromatography
RUO
Antibody
Monoclonal
ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
1G7
Unconjugated
Liquid
Mouse
IgG2a
100 μg
4°C or -80°C if preferred
Primary
Virus