Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C Virus Core Antigen Mouse anti-Virus, Clone: 1H7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA17611

Catalog No. 01-670-345


Add to cart

Description

Description

MA1-7611 detects HCV from virus samples. MA1-7611 has been successfully used in ELISA, Western blot and immunoprecipitation procedures. The MA1-7611 immunogen is a purified recombit HCV core 00115V protein.

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Specifications

Specifications

Hepatitis C Virus Core Antigen
Monoclonal
Unconjugated
Liquid
Mouse
IgG1
100 μg
4°C or -80°C if preferred
Primary
Virus
ELISA, Immunoprecipitation, Western Blot
1H7
PBS with 0.01% sodium azide
HepC; HepC core; HCV core
Purified recombit HCV core 00115V protein.
Protein A
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit