Learn More
Hepatitis C Virus Core Antigen Envelope Rabbit anti-Virus, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14114
Catalog No. PA14114
Add to cart
Description
Description
PA1-4114 detects Hepatitis C Virus. PA1-4114 has been successfully used in Western blot procedures. The PA1-4114 immunogen is a synthetic peptide corresponding to residues C(33) G V Y L L P R R G P R(43) of HCV core, envelope, and part of E2/NS1 of HCV.
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Specifications
Specifications
Hepatitis C Virus Core Antigen Envelope
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
HepC; HepC core; HCV core; HCV envelope; HepC envelope
Synthetic peptide corresponding to residues C(33) G V Y L L P R R G P R(43) of HCV core, env, and part of E2/NS1 of HCV.
Antigen affinity chromatography
RUO
Antibody
Polyclonal