Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF2 Rabbit anti-Human, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124459

Catalog No. PIPA124459


Add to cart

Description

Description

PA1-24459 detects Hepatocyte Nuclear Factor 1-beta (TCF2) in Human samples. PA1-24459 has been successfully used in Western Blot procedures. PA1-24459 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human TCF2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Hepatocyte Nuclear Factor 1? (HNF-1?, Homeoprotein LFB3, Transcription factor 2, TCF2, Variant hepatic nuclear factor ) is a liver-specific factor of the homeobox-containing basic helix-turn-helix family. The TCF2 protein is believed to form heterodimers with another liver-specific member of this transcription factor family, TCF1; depending on the TCF2 isoform, the result may be to activate or inhibit transcription of target genes. Mutation of TCF2 that disrupts normal function has been identified as the cause of MODY5 (Maturity-Onset of Diabetes, Type 5). A third human transcript variant is believed to exist based on such a variant in the rat: however, to date such an mRNA species has not been isolated.
Specifications

Specifications

TCF2
Polyclonal
Unconjugated
HNF1B
Liquid
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6928
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P35680
HNF1B
Synthetic peptide region of human TCF2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit