Learn More
Hepatocytes Mouse anti-Human, Clone: EP-1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135394
Catalog No. 01-671-449
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody has been successfully used in immunohistochemistry applications and stains human liver canaliculi and a subset of hepatocellular carcinomas.
Specifications
Specifications
Hepatocytes
Monoclonal
0.1 mg/mL
PBS with 0.05% sodium azide
Mouse
IgG2b
100 μg
4° C
Primary
Human
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
EP-1
Unconjugated
Liquid
Hepatocyte Specific Antigen
Purified
RUO
Antibody
Monoclonal