Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524340

Catalog No. PIPA524340


Add to cart

Specifications

Specifications

HEPN1
Polyclonal
Unconjugated
HEPN1
Liquid
HEPN1
KLH conjugated synthetic peptide between 6-34 amino acids from the N-terminal region of human HEPN1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q6WQI6
HEPACAM opposite strand 1, cancer susceptibility gene HEPN1, putative cancer susceptibility gene HEPN1 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
641654
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit