Learn More
HSV Type 1 Mouse anti-Virus, Clone: HP1.3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110679
Catalog No. 01-671-910
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10679 detects Herpes Simplex Virus, type 1 from infected samples. MA1-10679 has been successfully used in ELISA and immunofluorescence applications. The MA1-10679 immunogen is HSV I and II-infected cells.
HSV is a member of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer′s disease.
Specifications
Specifications
HSV Type 1
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
HSV1; Herpes Simplex I; HSV Type 1; Herpes Simplex 1; HHV; HSV; Herpes Simplex Type 1; KITH_HHV1S
HSV I and II-infected cells.
Purified
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence
HP1.3
Unconjugated
Liquid
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus