Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes Simplex Virus Type 1/2 Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA127209
Description
PA1-27209 will not react with HEp-2 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Herpes Simplex Virus Type 1/2 Polyclonal antibody specifically detects Herpes Simplex Virus Type 1/2 in Virus samples. It is validated for Immunocytochemistry.Specifications
Herpes Simplex Virus Type 1/2 | |
Polyclonal | |
FITC | |
Herpes Simplex 1; Herpes Simplex 2; Herpes Simplex I; Herpes Simplex II; HHV; HHV-2; HSV; HSV 1/2; HSV I/II; HSV Type 1; HSV Type 2; HSV Type II; HSV1; HSV2; HSV-2 | |
Human HSV1, strain F infected cell lysate. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction