Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSV Type 1, FITC, Polyclonal, Invitrogen™

Catalog No. PIPA129005
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA129005 100 μL
1 options

Catalog No. PIPA129005

Supplier: Thermo Scientific PA129005

Rabbit Polyclonal Antibody

Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer's disease.

PA1-29005 detects HSV1 (FITC ) from human samples.PA1-29005 has been successfully used in immunofluorescence applications.The PA1-29005 immunogen is: Herpes Simplex Virus (HSV), Strain F (human)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Simplex Virus Type 1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4mg/mL
Conjugate FITC
Gene Alias HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, KITH_HHV1S
Host Species Rabbit
Immunogen Herpes Simplex Virus (HSV), Strain F (human)
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.