Learn More
HSV Type 2 Mouse anti-Virus, Clone: HP105.4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110683
Catalog No. 01-671-914
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10683 detects Herpes Simplex Virus, type 2 from HSV II ICP6 samples. MA1-10683 has been successfully used in ELISA and immunofluorescence applications. The MA1-10683 immunogen is HSV II-infected cells.
Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer′s disease.
Specifications
Specifications
HSV Type 2
Monoclonal
Unconjugated
Liquid
Mouse
IgG2b
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus
ELISA, Immunofluorescence
HP105.4
PBS with no preservative; pH 7.4
HSV2; HSV-2; Human herpes virus 2; HHV-2; Herpes Simplex II; HSV Type 2; Herpes Simplex 2; HSV Type II
HSV I and II-infected cells.
Protein G
RUO
Antibody
Monoclonal