Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus Type 1/2 Rabbit anti-Virus, FITC, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA17215

Catalog No. PA17215


Add to cart

Description

Description

PA1-7215 detects the herpes simples virus. This antibody cross reacts with types 1 and 2. PA1-7215 has been successfully used in immunofluorescence procedures. The PA1-7215 immunogen is strain F (human).

Members of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer ft.s disease.
Specifications

Specifications

Herpes Simplex Virus Type 1/2
Polyclonal
FITC
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
Immunofluorescence
4-5 mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
HSV1; Herpes Simplex I; HSV Type 1; Herpes Simplex 1; HHV; HSV; Herpes Simplex Type 1; HSV2; HSV-2; Human herpes virus 2; HHV-2; Herpes Simplex II; HSV Type 2; Herpes Simplex 2; HSV Type II; HSV 1/2; HSV I/II
Strain F (human).
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit