Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hERV-WL Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC505
Description
Detect HERV using this rabbit polyclonal antibody, Anti-hERV-WL Antibody validated for use in western blotting and Flow Cytometry.
Specifications
hERV-WL | |
Polyclonal | |
Rabbit polyclonal serum in buffer containing 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Human | |
Serum |
Flow Cytometry, Western Blot | |
Unconjugated | |
Sec23b | |
Recombinant protein corresponding to human ERV-WL. | |
100 μL | |
n/a | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction