Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HES-1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130912

Catalog No. NBP130912


Add to cart

Description

Description

HES-1 Polyclonal antibody specifically detects HES-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

HES-1
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:2000, Immunoprecipitation 1:100-1:500
Affinity Purified
HES1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human HES1. The exact sequence is proprietary.
30 kDa
100 ul
Neuronal Stem Cell Markers, Stem Cell Markers
Primary
3280
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
BHLHB39, bHLHb39hairy homolog (Drosophila), Class B basic helix-loop-helix protein 39, FLJ20408, Hairy and enhancer of split 1, hairy and enhancer of split 1, (Drosophila), Hairy homolog, Hairy-like protein, Hes1, HES-1, hHL, HL, HRYHHL, transcription factor HES-1
Rabbit
IgG
Antigen Affinity-purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Predicted cross-reactivity based on sequence identity: Japanese Medaka (93%) and Xenopus tropicalis (93%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only