Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526717

Catalog No. PIPA526717

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HES7 is a member of the hairy and enhancer of split family of bHLH transcription factors. The protein functions as a transcriptional repressor, and is implicated in correct patterning of the axial skeleton.
Specifications

Specifications

HES7
Polyclonal
PBS with 0.09% sodium azide
Q9BYE0, Q8BKT2
SCDO4, bHLHb37, bHLH factor Hes7, class B basic helix-loop-helix protein 37, hHes7, transcription factor HES-7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84653, 84667
Western Blot
Unconjugated
HES7
Liquid
HES7
KLH conjugated synthetic peptide between 26-55 amino acids from the N-terminal region of human HES7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit