Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hexamethylene bis-acetamide inducible 1 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human HEXIM1 protein.

Manufacturer:  Abnova Corporation H00010614B02

Catalog No. 89-007-023


Add to cart

Description

Description

Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. This gene has no introns. [provided by RefSeq

Sequence: MAEPFLSEYQHQPQTSNCTGAAAVQEELNPERPPGAEERVPEEDSRWQSRAFPQLGGRPGPEGEGSLESQPPPLQTQACPESSCLREGEKGQNGDDSSAGGDFPPPAEVEPTPEAELLAQPCHDSEASKLGAPAAGGEEEWGQQQRQLGKKKHRRRPSKKKRHWKPYYKLTWEEKKKFDEKQSLRASRIRAEMFAKGQPVAPYNTTQFLMDDHDQEEPDLKTGLYSKRAAAKSDDTSDDDFMEEGGEEDGGSDGMGGDGSEFLQRDFSETYERYHTESLQNMSKQELIKEYLELEKCLSRMEDENNRLRLESKRLGGDDARVRELELELDRLRAENLQLLTENELHRQQERAPLSKFGD
Specifications

Specifications

hexamethylene bis-acetamide inducible 1
Unconjugated
No additive
NM_006460.2
CLP1|EDG1|FLJ13562|HIS1|MAQ1
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human HEXIM1 protein.
HEXIM1
Serum
HEXIM1
HEXIM1 (NP_006451.1, 1 a.a. ~ 359 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
10614
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit