Learn More
HK2 Recombinant Rabbit Monoclonal Antibody (2H8L6), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific 700422
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombit rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombit rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
In vertebrates there are four major glucose-phosphorylating isoenzymes, designated hexokinase I, II, III, and IV. Hexokinase activity is involved in the first step in several metabolic pathways including phosphorylation of glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Hexokinase is an allosteric enzyme inhibited by its product GLC-6-P. Hexokinase 2 is the predomit hexokinase isozyme expressed in insulin-responsive tissues such as skeletal muscle. Expression of this gene is insulin-responsive, and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells.Specifications
HK2 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P52789 | |
HK2 | |
A recombit protein corresponding to amino acids 325-444. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
2H8L6 | |
Unconjugated | |
HK2 | |
HK II, hexokinase type II, hexokinase-2, hexokinase-2 muscle, muscle form hexokinase | |
Rabbit | |
Protein A | |
RUO | |
3099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.