Learn More
HFH-4/FOXJ1 Mouse anti-Human, Mouse, Rat, Clone: 3-19, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05837
Catalog No. 05-837-MI
Add to cart
Description
Description
Anti-HFH-4/FOXJ1 Antibody, clone 3-19 is a high quality Mouse Monoclonal Antibody for the detection of HFH-4/FOXJ1 and has been validated in western blotting and immunohistochemistry.
Specifications
Specifications
HFH-4/FOXJ1
3-19
NM_053832
FOXJ1, HFH-4, HFH4, MGC35202, FKHL13
Full length rat hepatocyte nuclear factor homolog (HFH-4)/forkhead box protein J1 (FOXJ1) GST fusion protein
Ascites
−20°C for up to 24 months.
Monoclonal
Immunohistochemistry, Western Blot
Unconjugated
Ascites
Murine
κ
200μL
Primary
Human, Murine, Rat