Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HFM1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524112

Catalog No. PIPA524112

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HFM1
Polyclonal
PBS with 0.09% sodium azide
A2PYH4
MER3, RP11-539G11.1, SEC63D1, Si-11, Si-11-6, helicase, SEC63 domain containing 1, SEC63 domain-containing protein 1, helicase-like protein HFM1, probable ATP-dependent DNA helicase HFM1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
164045
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HFM1
Liquid
HFM1
KLH conjugated synthetic peptide between 1293-1321 amino acids from the C-terminal region of human HFM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit