Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGD Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP155134

Catalog No. NBP155134


Add to cart

Description

Description

HGD Polyclonal antibody specifically detects HGD in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

HGD
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
HGD
Synthetic peptides corresponding to HGD(homogentisate 1,2-dioxygenase (homogentisate oxidase)) The peptide sequence was selected from the middle region of HGD. Peptide sequence KLFAAKQDVSPFNVVAWHGNYTPYKYNLKNFMVINSVAFDHADPSIFTVL.
50 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:100-1:2000
Q93099
AKU, EC 1.13.11.5, HGOFLJ94126, homogentisate 1,2-dioxygenase, homogentisate 1,2-dioxygenase (homogentisate oxidase), homogentisate oxidase, Homogentisate oxygenase, Homogentisic acid oxidase, homogentisicase
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
3081
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only