Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hedgehog interacting protein, Mouse, Clone: 5D11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HHIP.
Supplier: Abnova Corporation H00064399M01
Description
This gene encodes a protein similar to the mouse hedgehog-interacting protein, a regulatory component of the hedgehog signalling pathway. Members of the hedgehog family are evolutionarily conserved proteins which are involved in many fundamental processes in embryonic development, including anteroposterior patterns of limbs and regulation of left-right asymmetry. [provided by RefSeq
Sequence: GDAKFGERNEGSGARRRRCLNGNPPKRLKRRDRRMMSQLELLSGGEMLCGGFYPRLSCCLRSDSPGLGRLENKIFSVTNNTECGKLLEEIKCALCSPHSQSpecifications
hedgehog interacting protein | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_022475 | |
HHIP | |
HHIP (NP_071920, 21 a.a. ∼ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
5D11 | |
Mouse monoclonal antibody raised against a partial recombinant HHIP. | |
HHIP | |
FLJ20992/FLJ90230/HIP/STQTL12 | |
Mouse | |
Affinity Purified | |
RUO | |
64399 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction