Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

HIAT1 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526974

Catalog No. PIPA526974

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HIAT1
Polyclonal
PBS with 0.09% sodium azide
Q96MC6, P70187
hippocampus abundant gene transcript 1, hippocampus abundant transcript 1 protein, putative tetracycline transporter-like protein, tetracycline transporter-like protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100772600, 15247, 64645
Immunohistochemistry, Western Blot
Unconjugated
MFSD14A
Liquid
Mfsd14a
KLH conjugated synthetic peptide between 454-483 amino acids from the C-terminal region of human HIAT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit